Tlačiť
ASTM colour reference standards, ASTM D 6045, D1500, Supelco®
Dodávateľ: Merck
SIAL134010-500MLEA 0 EUR
SIAL134010-500ML SIAL134030-500ML SIAL134020-500ML SIAL134000-500ML SIAL134290-500ML
ASTM colour reference standards, ASTM D 6045, D1500, Supelco®
ASTM Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to International Standards
  • 500mL Pack Size supplied with MSDS
  • 18 months shelf life
  • Tamper evident security packaging