Tlačiť
Precision laboratory thermometers with safety-coating, ACCU-SAFE, stem type
Dodávateľ: Ludwig Schneider
Accu-Safe
SCNL1132006S/80EA 0 EUR
SCNL1132006S/80 SCNL1052303S/80 SCNL1042376S/80 SCNL1042380S/80 SCNL1042373S/80 SCNL1042384S/80 SCNL1132005S/80 SCNL1052105S/80 SCNL1042375S/80 SCNL1132007S/80 SCNL1042377S/80 SCNL1042374S/80 SCNL1042379S/80 SCNL1132004S/80
Precision laboratory thermometers with safety-coating, ACCU-SAFE, stem type
Teploměry Analogue Thermometers
These ACCU-SAFE thermometers measures precisely and reliably without mercury from the very start.
  • Round, white back capillary form
  • High transparency for easy legibility
  • Usable as stirring thermometer
  • Suitable for calibration
The rugged encapsulation protects against splintered glass or display-fluid leakage. The protective coating is remarkable for its high transparency and quick response time allowing easily legibility. It provides temperature resistance (up to 250 °C) and hence sterilisability, and a high degree of acid and alkali resistance (hydrofluoric acid).
Informace o dodávce: With individual serial number and certificate of conformity.
Balenie: Each item packed in square plastic sleeve.