Tlačiť
Muffle furnaces with lift up door, Tmax 1100 °C and controller C450, LT 3/11 - LT 40/11
Dodávateľ: Nabertherm
NABEL-152H1TN1EA 5151.2 EUR
NABEL-152H1TN1 NABEL-152H1TN1T NABEL-402H1TN1GT NABEL-052H1TN1Z NABEL-092H1TN1 NABEL-402H1TN1 NABEL-242H1TN1Z NABEL-402H1TN1G NABEL-242H1TN1TZ NABEL-032H1TN1GT NABEL-152H1TN1TZ NABEL-092H1TN1G NABEL-052H1TN1GT NABEL-092H1TN1GT NABEL-032H1TN1T NABEL-092H1TN1T NABEL-092H1TN1TZ NABEL-032H1TN1 NABEL-242H1TN1T NABEL-402H1TN1TZ 455-0657 NABEL-032H1TN1G NABEL-052H1TN1T NABEL-032H1TN1Z NABEL-402H1TN1T NABEL-152H1TN1G NABEL-152H1TN1Z NABEL-242H1TN1GT NABEL-402H1TN1Z NABEL-052H1TN1G NABEL-242H1TN1G NABEL-052H1TN1 NABEL-092H1TN1Z NABEL-152H1TN1GT NABEL-052H1TN1TZ NABEL-032H1TN1TZ
Muffle furnaces with lift up door, Tmax 1100 °C and controller C450, LT 3/11 - LT 40/11
Sušárny Pece vysokoteplotní
The LT 3/11 - LT 40/11 muffle furnaces are the ideal choice for daily laboratory use. These high quality models have an advanced and attractive design, and are very reliable. The muffle furnaces have a lift up door as standard.
  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Lift up door (LT) with hot surface facing away from the operator
  • Exhaust air outlet on the rear wall of furnace
  • Solid state relays provide low-noise operation
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: Recording of process data with USB-flash drive
Heating is from two sides by means of ceramic heating plates (heating from three sides for muffle furnaces LT 24/11 - LT 40/11). These ceramic heating plates with an integral heating element are protected from fumes and splashing, and are easy to replace. Only fibre materials which are not classified as carcinogenic according to TRGS 905, class 1 or 2 are used.
The furnace is equipped with C450 professional controller with LC display, featuring 10 freely adjustable programs and 20 segments and two extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The C450 controllers can automatically activate the vent with fan or catalyst (not model LT 15).
Objednávacie informácie: All units require a 230 V single-phase power supply, except LT 24 and LT 40 (400 V 3/N/PE). Optional accessories e.g. a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of furnace. Manual or automatic gas supply systems are also available.
Pozor: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model.
Note heat up times: If connected at 230 V 1/N/PE rsp. 400 V 3/N/PE.
Outer dimensions include opened lift up door.
* Temperature uniformity of ± 5K in the empty work space for temperatures >800 °C.