Tlačiť
Muffle furnaces with fold down door, Tmax 1200 °C and standard controller B510, L 3/12 - L 40/12
Dodávateľ: Nabertherm
Nabertherm
455-1537EA 7997.12 EUR
455-1537 455-1834 455-1536 455-1833 455-1734 455-1836 455-1739 455-1023 455-1022 455-1025 455-1124 455-1740 455-1123 455-1742 455-1544 455-1543 455-1326 455-1546 455-1325 455-1327 NABEL-054K2LN2TZ 455-1827 455-1826 455-1828 455-1333 455-1332 455-1016 455-1115 455-1335 455-1114 455-1015 455-1535 455-1733 455-1017 455-1732 455-1116
Muffle furnaces with fold down door, Tmax 1200 °C and standard controller B510, L 3/12 - L 40/12
Sušárny Pece vysokoteplotní
The L 3/12 - L 40/12 muffle furnaces are the ideal choice for daily laboratory use. These high quality models have an advanced and attractive design, and are very reliable. The muffle furnaces have a fold down door.
  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Fold down door (L models) can be used as a work platform
  • Exhaust air outlet on the rear wall of furnace
  • Solid state relays provide low-noise operation
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: Recording of process data with USB-flash drive
Heating is from two sides by means of ceramic heating plates (heating from three sides for muffle furnaces L 24/12 - L 40/12). These ceramic heating plates with an integral heating element are protected from fumes and splashing, and are easy to replace. Only fibre materials which are not classified as carcinogenic according to TRGS 905, class 1 or 2 are used.
The furnace is equipped with a B510 standard controller with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation. The B510 controllers can automatically activate the vent with fan or catalyst (not model L 15).
Objednávacie informácie: All units require a 230 V single-phase power supply, except L 24 and L 40 (400 V 3/N/PE). Optional accessories e.g. a vent, vent with fan or catalytic converter, over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, protective gas connection on the rear wall of the furnace. Manual or automatic gas supply systems are also available. Corresponding models with lift up doors, different Tmax and other controllers are also available. Please search the website for more details.
Pozor: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model.
Note heat up times: If connected at 230 V 1/N/PE rsp. 400 V 3/N/PE.
* Temperature uniformity of ± 5K in the empty work space for temperatures >800 °C.