Tlačiť
Capillary viscometers, Visco.Fix system
Dodávateľ: Lauda
LAUDEGVH704EA 0 EUR
LAUDEGVH704 LAUDEGVH703 LAUDEGVZ001
Capillary viscometers, Visco.Fix system
Viskozimetry Viskozimetry kapilární
The standard-compliant Ubbelohde and Micro-Ubbelohde capillary tubes with Visco.Fix are effectively protected. Significantly longer lifetime due to made-to-last design. Higher precision due to longer use of the same capillary. Always the right capillary at hand with the Visco.Fix carousel.
  • Easy cleaning with individual beakers
  • For clean and safe draining and drying
  • No more dust inside your capillaries
Capillary tubes for viscosities from 0,3 to 30000 mm/s, also available as dilution capillary or for automatic cleaning, with calibration certificate, durable ring marks and serial numbers.
Informace o dodávce: Visco.Fix package consists of Visco.Fix viscometer (EGVH 703 and EGVH 704) two each and one carousel.