Tlačiť
Giemsa's stain solution, Gurr™ for microscopical staining
Dodávateľ: VWR Chemicals
352603REA 90.23 EUR
352603R
Giemsa's stain solution, Gurr™ for microscopical staining
Giemsa's stain
Číslo MDL: MFCD00081642
Číslo CAS: 51811-82-6
IVDLogo
Order Now
Výsledky špecifikačného testu
Suitability for microscopy (blood smear) Passes test
Erythrocytes Beige-brownish
Nuclei Violet
Eosinophilic granules Reddish to red-brown
Neutrophilic granules Red-violet
Lymphocyte cytoplasm Blue
Monocyte plasma Grey-blue
Thrombocytes Violet
Basophilic granules Dark violet
Blood parasite nuclei Light red