Giemsa's stain solution, Gurr™ for microscopical staining
Dodávateľ: VWR Chemicals
352603REA
90.23
EUR
352603R
Giemsa's stain solution, Gurr™ for microscopical staining
Giemsa's stain
Číslo MDL:
MFCD00081642 Číslo CAS: 51811-82-6 |
![]() |
Výsledky špecifikačného testu
Suitability for microscopy (blood smear) | Passes test |
Erythrocytes | Beige-brownish |
Nuclei | Violet |
Eosinophilic granules | Reddish to red-brown |
Neutrophilic granules | Red-violet |
Lymphocyte cytoplasm | Blue |
Monocyte plasma | Grey-blue |
Thrombocytes | Violet |
Basophilic granules | Dark violet |
Blood parasite nuclei | Light red |