Lugol's iodine solution, Gurr™ for microscopy
351903YEA
53.63
EUR
351903Y
Lugol's iodine solution, Gurr™ for microscopy
Lugol's iodine solution
![]() |
Výsledky špecifikačného testu
Suitability for microscopy | Passes test |
Staphylococcus aureus | Dark bluish violet |
Escherichia coli | Pink to red |