Tlačiť
Pesticide reference standard, Penflufen, PESTANAL®, analytical standard, Supelco®
Dodávateľ: Merck
PESTANAL®
37049-100MGEA 454.46 EUR
37049-100MG
Pesticide reference standard, Penflufen, PESTANAL®, analytical standard, Supelco®
Standardy Standardy organické
Penflufen might be used as a reference standard in the determination of penflufen in vegetables and cereals using QuEChERS technique followed by ultra high-performance liquid chromatography coupled with tandem mass spectrometry (UHPLC−MS/MS).
  • It is a member of pyrazole amide fungicide
  • Helps in controlling fungal diseases in crop fields