Tlačiť
Pesticide reference standard, Penoxsulam, PESTANAL®, analytical standard, Supelco®
Dodávateľ: Merck
PESTANAL®
32094-25MGEA 99.98 EUR
32094-25MG
Pesticide reference standard, Penoxsulam, PESTANAL®, analytical standard, Supelco®
Standardy Standardy organické
Penoxsulam may be used as a reference standard in the determination of penoxsulam in water samples using high performance liquid chromatography coupled with diode array detector (HPLC-DAD), aquatic systems adjacent to rice fields using liquid chromatography coupled with ultraviolet detector.
  • It is a triazolopyrimidine sulphonamide type of post-emergence herbicide
  • Utilised as an acetolactate synthase (ALS) inhibitor for weed control