Tlačiť
Gas mixtures for GC Calibration, analytical standards, Supelco®
Dodávateľ: Merck
501654.EA 0 EUR
501654. 23462. 23463. 501662.
Gas mixtures for GC Calibration, analytical standards, Supelco®
Standardy Standardy pro environmentální analýzu
Multi-component gas mixture suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analysers.
  • A certificate of analysis is included
  • Packaged in lightweight, non-refillable, recyclable containers
  • Portable, convenient, and easy to use
  • Manufactured gravimetrically for maximum accuracy
  • NIST traceable