Tlačiť
RALA320300
720-0194EA 63.71 EUR
720-0194
RALA320300
Giemsa Staining in Parasitology and Mycology
Giemsa technique is the most useful panoptic staining method in parasitology, namely in the field of tissular and blood protozoa. It can be used in medical and veterinary mycology. Giemsa stains cytoplasmic structures blue, and nuclear structures or other DNA-containing organites (like the kinetoplast of trypanosomatids) purple red.