Tlačiť
HPLC columns, Exsil™ SCX
Dodávateľ: Hichrom Limited
Exsil™
EXSCX-5-50AEA 0 EUR
EXSCX-5-50A EXSCX-5-100A EXSCX-5-125A EXSCX-5-10C5 EXSCX-5-150A EXSCX-5-125AF EXSCX-5-100AS EXSCX-5-250A EXSCX-5-250AF
HPLC columns, Exsil™ SCX
Kolony chromatografické
Exsil™ HPLC columns are a useful alternative to the classical spherical HPLC silicas.
  • Spherical porous silica
  • Competitively priced
  • High efficiency
Exsil™ 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications. Exsil™ 80 is a higher surface area, complementary range of materials.