Tlačiť
Ice and water dispenser
Dodávateľ: Scotsman
474-0017
Ice and water dispenser
Výrobníky ledu
Makes and dispenses soft, chewable nugget ice.
  • Flexible utility location
Features include sleek, european design with attractive rounded corners and informative instruction labels, up to 25% less fan noise allows installation of the unit in the quietest hallways, improved dispensing with easier access and room for most containers.
TouchFree® actuation provides convenient one-hand operations and eliminates the risk of cross contamination.
Informace o dodávce: All units are delivered without a plug.