Tlačiť

Dimethylsulfoxid ≥99.9%, AcroSeal™ for HPLC


Packaged under argon.

47330.K2EA 168.12 EUR
47330.K2
Dimethylsulfoxid ≥99.9%, AcroSeal™ for HPLC
Dimethylsulfoxid

Packaged under argon in resealable AcroSeal® bottles.

Vzorec: (CH₃)₂SO
Mw: 78,14 g/mol
Teplota varu: 189 °C
Teplota tavenia: 18,4 °C
Hustota: 1,101
Teplota vzplanutia: 87 °C (189 °F)
Skladovacia teplota: Ambient
Číslo MDL: MFCD00002089
Číslo CAS: 67-68-5
EINECS: 200-664-3
https://sk.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR