Tlačiť
Trypsin, from Bovine Pancreas
VWRC39041CU VWRC390414M
Trypsin, from Bovine Pancreas
(formerly EC 3.4.4.4)
Contains about 85% lactose as diluent; activity about 0.5 Anson units per g (calculated as at 35.5°C). 1 Anson unit is that which hydrolyses urea denatured haemoglobin at such an initial rate under the specified conditions (25°C) that the hydrolysis products formed per minute give the same absorbance with the Folin and Ciocalteu's reagent as 1 millimole of tyrosine. (Based on Anson, M.L., J. Gen. Physiol., 1938, 22, 79)
For in-vitro laboratory use only.