Tlačiť
Acrylogel 2.6 solution 30%, Electran for electrophoresis, VWR®
427205EEA 0 EUR
VWRC427205E
Acrylogel 2.6 solution 30%, Electran for electrophoresis, VWR®
A convenient, ready to use 30% w/v (30%T) solution of acrylamide `Electran` and NN'-methylenebisacrylamide `Electran` (2.6% C) in deionised water.
(Final ratio is 37.5:1)