Tlačiť
Aqua textile drop down mats 45, Nomad™
Dodávateľ: 3M
3M™
MMMA45SZ69EA 0 EUR
MMMA45SZ69 MMMA45BN69 MMMA45SZ1218 MMMA45SZ915 MMMA45BU915
Aqua textile drop down mats 45, Nomad™
Rohože Rohože podlahové, permanentní
3M™ Nomad™ aqua textile drop down matting series 45 are a high performance entrance mats for low traffic applications.
  • Entrance mat for low traffic applications (up to 500 crossings per day)
  • Backed with slip resistant (DIN 51130: R11) PVC
  • Can be easily bonded and edged
  • Edge sealing 25 mm
  • EN 13501-1: Efl-s1 fire resistance
  • Number of loops 85680/m²
The true cost of entrance matting is not what you pay for the product but what it can save you in annual cleaning and maintenance costs over its lifetime. 3M™ Nomad™ aqua textile entrance matting gives you longer lasting floor protection at a lower total cost, by stopping up to 80% of dirt and moisture at the door. Dual-fibre tuft traps and absorbs dirt and moisture more effectively than conventional matting.