Tlačiť
Industrial label printers, BradyPrinter i5100, standard
Dodávateľ: Brady
BRDY149461EA 2741.32 EUR
BRDY149461 BRDY149458 BRDY198778
Industrial label printers, BradyPrinter i5100, standard
Tiskárny Label Printers
The BradyPrinter i5100 industrial label printer offers a perfect mix of performance and smarts.
  • Asset identification, circuit board and component identification
  • General and industrial identification
This benchtop printer can print on 70 label materials, more than 1200 stock label parts and tailored custom label materials and parts. Applications include a wide range of specialised and commonly used product identification labels, electronics and PCB labels, logistics and asset tracking labels, cable and component identification labels and laboratory sample labels.
Mono colour. High quality printing on reliable, industrial grade labels from 5 up to 110 mm. Driven by a powerful 32 bit and 800 Mhz internal printer processor. Prints at a speed of up to 300 mm/s at 300 dpi.
Certifikácie: FCC Class A, cUL.
Objednávacie informácie: Media and ribbon, supplied separately.
Informace o dodávce: This i5100 printer comes in a cardboard box. It ships with a power cord, a USB cable, a product CD holding the driver, the user manual in languages, the configuration/settings manual in English and a printed user manual in English, an RFID enabled roll holder, a ribbon take up spindle and a Brady
Workstation Basic Suite or a Brady Workstation PWID Suite.