Tlačiť
Polariser set for ring light, VisiLED, S80-65N
Dodávateľ: Schott AG
SCOC400665EA 1004.62 EUR
SCOC400665
Polariser set for ring light, VisiLED, S80-65N
Polarisation system for brightfield ringlights. Turnable polariser with adapting thread and sliding filter as analyser.
  • Polarised light by a polarising and analysing filter within the beam path can be very efficiently used in stereo microscopy
  • Use to contrast inner material structure or to identify metal particles within others
Designed for use only with VisiLED ringlight S80-65N.
Objednávacie informácie: VisiLED ringlight and controller MC 750, MC 1100, or MC 1500 required.