Tlačiť
Lumitester™ Smart
Dodávateľ: HyServe
SCIL61234EA 2977.08 EUR
SCIL61234
Lumitester™ Smart
The Lumitester™ Smart offers high sensitivity detection and wireless connection capabilities.
  • Easy-to-use, results within 10 seconds
  • App to continuously monitor multiple data
  • Centralized multi-site data with cloud data management
Lumitester™ Smart used with LuciPac Pen is a quick and simple device for hygiene control in ten seconds.
The LuciPac™ pen has the advantage that it not only measures ATP, but AMP as well. AMP is a degradation product of ATP and can be found in food residues and other biological dirt. The kit is recommended for applications where the user has to be mobile and wants a quick and easy-to-use tool for hygiene control.
This is a new hygiene control system based on the luciferase reaction. The system includes LuciPac™ A3 Surface, LuciPac™ A3 Water and Lumitester™ Smart. The Lumitester™ Smart replaces Lumitester™ PD-30, which is no longer produced. Lumitester™ Smart has Bluetooth and can transfer data to your mobile or tablet or via a cable to your PC. There is also a cloud app for storing and presenting the data. LuciPac™ A3 measures ATP+ADP+AMP, while other luciferase-based hygiene measures only ATP. Enzymes and heat degrade ATP to ADP and AMP.
Informace o dodávce: Supplied with 2 size AA alkaline batteries, cleaning brush, USB cable, strap, and quick manual.