Tlačiť
Liquid detergents for automated dishwashing, concentrated, neodisher® Alka 240
Dodávateľ: Dr. Weigert
neodisher®
WEIG311113EA 0 EUR
WEIG311113 WEIG311126
Liquid detergents for automated dishwashing, concentrated, neodisher® Alka 240
Prostředky čistící Detergenty a povrchové čističe
The special phosphate-free and active chlorinefree formula enables excellent dispersion of food residues and removal of residues of tanning agents. Excellent starch dissolving and complexing action.
  • Viscosity: <50 mPas (concentrate, 20 °C)
  • Density: 1,3 g/cm³ (20 °C)
  • Conductivity: 6,2 mS/cm (3 g/L, determined in deionised water, 60 °C)
  • Titration factor: 0,43
It is a dried-on starch-containing residues and starch and protein deposits which is suitable for cleaning items with slightly dried-on
coffee and tea residues especially suitable for use with soft to medium hard water. It is suitable for glass, porcelain, stainless steel and alkali-compatible synthetic materials but not suitable for aluminium, anodised aluminium and light metal alloys.
Fields of application: Cleaning of dishes, glasses, cutlery, trays and working utensils in professional dishwashers in all food-processing companies, such as professional kitchens, butcher's shops and bakeries.
Storage and transport is only permitted in original packaging. The solution has to be rinsed off completely with water. Rinse out dosing system including suction hose with water before changing product. For economical and controlled dosing we recommend using automatic dosing devices or systems.
The temperature in the main washing tank should be at least 60 °C to ensure sufficient hygiene safety. When working with hard or salty water a special water treatment is recommended to achieve optimal results.
Contains <5% amphoteric surfactants, phosphonates, polycarboxylates according to regulation (EC) No 648/2004 on detergents.
Pozor: For professional use only. Do not mix with other products.