Tlačiť
Detergents for automated reprocessing of thermostable instruments, liquid concentrate, neodisher® DuoClean
Dodávateľ: Dr. Weigert
neodisher®
148-0154EA 88.24 EUR
148-0154 WEIG403026
Detergents for automated reprocessing of thermostable instruments, liquid concentrate, neodisher® DuoClean
Prostředky čistící Detergenty a povrchové čističe
Highly alkaline, surfactant-containing intensive detergent with good cleaning action for removing blood, protein and stubborn OR residues.
  • Can be used with water of any hardness
  • Even after a long drying time residues are removed safely
  • Free of phosphates, silicates and oxidising agents
  • Viscosity: <50 mPas (concentrate, 20 °C)
  • Density: 1,2 g/cm³ (20 °C)
  • Titration factor: 0,55
Use suitable dosing devices. Rinse out dosing system including suction hose with water before changing product. Reprocessing should comply with all ordinances pursuant to the regulations on medical devices and should be performed with appropriate validated processes.
Suitable for medical instruments made of stainless steel and other alkali-compatible utensils made of glass and plastics but not suitable for cleaning utensils made of aluminium and anodised aluminium. Colour changes might occur with titanium alloys. This must be taken into account with regard to colour-coded implants. Badly soiled gynaecological and orthopaedic instruments as well as instruments for high-frequency surgery can be reprocessed in the two step neodisher® DuoClean process without laborious pre- or after-treatment.
Contains <5% amphoteric surfactants, 5 to 15% polycarboxylates according to regulation (EC) No 648/2004 on detergents.
Pozor: For professional use only. Do not mix with other products.