Tlačiť

Detergent for manual cleaning, liquid concentrate, neodisher® Septo PreClean

Dodávateľ: Dr. Weigert
148-0166EA 0 EUR
WEIG450335
Detergent for manual cleaning, liquid concentrate, neodisher® Septo PreClean
Cleaning and disinfecting agent for the manual reprocessing of thermostable and thermolabile instruments.

  • Excellent cleaning performance, non-proteinfixing
  • Excellent material compatibility
  • Free of aldehydes and quaternary ammonium compounds
  • Viscosity: <50 mPas (concentrate, 20 °C)
  • Density: 1,0 g/cm³ (20 °C)

Disinfecting pre-cleaning of thermostable and thermolabile instruments, dental instruments and flexible endoscopes example prior to automated reprocessing for optimum personnel safety. Wet disposal of surgical instruments and dental instruments over longer periods of time, example overnight or over the weekend. Also suitable for the manual cleaning and disinfection of thermostable and thermolabile instruments, including flexible endoscopes.

Suitable for stainless steel, anodised aluminium, non-ferrous metals (copper and brass), silicone, plastics. VAH¹-listed, on the IHO⁴-disinfectant list. neodisher® Septo PreClean is used in immersion and ultrasonic baths. Depending on the desired activity the working solution must be prepared according to the dosing recommendations. Place the instruments resp. endoscopes without air bubbles into the working solution and adhere to the below-mentioned contact times.

Contains <5% non-ionic surfactants, 5 to 15 % amphoteric surfactants also disinfectants, perfumes (limonene) according to regulation (EC) No 648/2004 on detergents.

Certifikácie: Bactericidal and yeasticidal activity according to VAH¹ methods and European standards, also active against enveloped viruses according to European standards and RKI/DVV³. Confirmed by certification.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR