Tlačiť
Antibacterial hand cleanser, Luxury Foam, Scott*
Dodávateľ: Kimberly-Clark
Scott® Scott
KIMB6348EA 323.34 EUR
KIMB6348
Antibacterial hand cleanser, Luxury Foam, Scott*
Péče o pokožku Mýdla a čističe pokožky
This antibacterial foam hand cleansers are effective against a range of micro-organisms.
  • Clear and frangrance free
  • Less water and time useage
The foam lather helps to wash hands thoroughly, but rinses off more quickly than traditional soap resulting in less water usage and time spent per wash. This hygienic hand wash, which is completely Triclosan Free, improves personal hygiene, cares for skin and reduces bacteria.
Ideal for use in busy washrooms in hygiene-critical areas such as healthcare, food processing and food service environments. Dispenses the ideal amount of soap, preventing overuse and delivers up to twice as many washes per litre as conventional liquid soaps; keeping costs down in areas of heavy use and reducing waste.
The cassettes are quick and easy to install with a simple push and click. When the cassette is empty, it can be compressed to conserve space and is fully recyclable once the pump is removed. Compatible with our Aquarius™® Hand Cleanser dispensers.
Certifikácie: Compliant to EN 1499.
Pozor: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904