Tlačiť
COVID-19 Antigen detection kit (SARS-CoV-2 N-protein detection), DNA DIAGNOSTIC
Dodávateľ: DNA Diagnostic
CV19ICEA 135.15 EUR
CV19IC
COVID-19 Antigen detection kit (SARS-CoV-2 N-protein detection), DNA DIAGNOSTIC
Systémy pro klinickou diagnostiku Clinical Reagents and Kits
The kit is a 15 minute chromatographic immunoassay for the qualitative detection of nucleoprotein (N) antigen of SARS-CoV-2 from nasal swab samples from patients suspected of COVID-19 infection.
The COVID-19 antigen detection kit is designed to detect the SARS-CoV-2 virus, which is responsible for the disease COVID-19.The virus group Coronavirus (Coronavirus, CoV) are sing-le-stranded, positive-stranded RNA virus. In addition to the new coronavirus that is causing COVID-19, a total of six coronaviruses are found to infect humans (HCoV-229E, HCoV-OC43, SARS-CoV, HCoV-NL63, HCoV-HKU1, and MERS-CoV)[1-3].The swab is used to collect an upper respiratory tract sample. The content of the Buffer ampoule is transferred to an extraction tube, which is used for the elution of the swab sample. After addition of a Drip Lid, three (3) drops of sample is added to the Detection Card.
The Detection Card pad is coated with an anti-SARS-CoV-2 N-Protein monoclonal IgY antibody which is labeled with colloidal gold. A second anti-SARS-CoV-2 N-Protein antibody is coated on the nitrocellulose membrane to make the Test line (T), and an anti-IgY secondary antibody is coated on the same nitrocellulose membrane to make a control line 'C'. The anti-SARS-CoV-2 N protein antibody reacts with the virus antigen (if present) and carry out chromatography along the nitrocellulose membrane, to react with the test line and control line, respectively. If the test result is valid, the control line is visual.
Balenie: Each box contains 25 tests.
Pozor: The test is intended for professional use only - intended users are trained personnel instructed in the in vitro diagnostic procedure.