Tlačiť
Yeast alpha-glucosidase
Dodávateľ: Apollo
APOSBIB3066-100KUEA 653.53 EUR
APOSBIB3066-100KU
Yeast alpha-glucosidase
Enzymy
From yeast (Saccharomyces cerevisiae). Specific activity: IU/mg protein>50 units. IU per 1mg powder is approximately 70 units.