Tlačiť
Yeast hexokinase
Dodávateľ: Apollo Scientific
APOSBIB3076-500UEA 73.69 EUR
APOSBIB3076-500U
Yeast hexokinase
Enzymy
Lyophilized powder. From yeast (Saccharomyces cerevisiae). Specific activity: IU/mg protein>140 units. IU per 1mg powder is approximately 100 units.