Tlačiť
Phosphogluconate dehydrogenase (decarboxylating)
Dodávateľ: Apollo Scientific
APOSBIB3094-25UEA 0 EUR
APOSBIB3094-25U
Phosphogluconate dehydrogenase (decarboxylating)
Enzymy
From Leuconostoc mesenteroides. Specific activity: IU/mg protein>30units. IU per 1ml suspension is approximately 600 units.
  • Ammonium sulfate suspension