Tlačiť
Recombinant keratinase
Dodávateľ: Apollo Scientific
APOSBIPR1000-1000UEA 0 EUR
APOSBIPR1000-1000U APOSBIPR1000-500U
Recombinant keratinase
Enzymy
Activity > 1400/U/mg. From Bacillus licheniformis and produced in E. Coli and B. Megaterium. Endotoxin negative. Sterile lyophilized powder. Chromatographically purified.