Tlačiť

Recombinant Pz-Peptidase

Dodávateľ: Apollo
APOSBIPR1006-300UEA 639.98 EUR
APOSBIPR1006-300U APOSBIPR1006-100U
Recombinant Pz-Peptidase
Enzymy
Activity >85 CDU/mg. From Bacillus licheniformis and produced in E.coli. Endotoxin negative. Sterile lyophilized powder.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR