Tlačiť
Bacillus halodurans alpha-Galactosidase 4A
Dodávateľ: Apollo
APOSBIZY1000-1MGEA 260.92 EUR
APOSBIZY1000-1MG
Bacillus halodurans alpha-Galactosidase 4A
Enzymy
alpha-Galactosidases are enzymes that participate in the hydrolysis of terminal, non-reducing alpha-D-galactose. Family: GH4. E.C.:3.2.1.22. Architecture: GH4. Main activity:alpha-Galactosidase.