Tlačiť
Clostridium cellulolyticum alpha-Galactosidase 27A
Dodávateľ: Apollo Scientific
APOSBIZY1003-1MGEA 0 EUR
APOSBIZY1003-1MG
Clostridium cellulolyticum alpha-Galactosidase 27A
Enzymy
alpha-Galactosidases are enzymes that participate in the hydrolysis of terminal, non-reducing alpha-D-galactose. Family: GH27. E.C.:3.2.1.22. Architecture: GH27. Main activity:alpha-Galactosidase.