Tlačiť
Clostridium perfringens N-Acetylglucosamine endo-beta-galactosidase 16C
Dodávateľ: Apollo Scientific
APOSBIZY1013-1MGEA 0 EUR
APOSBIZY1013-1MG
Clostridium perfringens N-Acetylglucosamine endo-beta-galactosidase 16C
Enzymy
beta-Galactosidases are enzymes that specifically hydrolyse terminal non-reducing beta-D-galactose residues in beta-Dgalactosides. Family: GH16. E.C.:3.2.1.-. Architecture: GH16. Main Activity:-acetylglucosamine endo-beta-galactosidase.