Tlačiť
Bacillus cereus Oligo-1,6-alpha-Glucosidase 13A
Dodávateľ: Apollo Scientific
APOSBIZY1029-1MGEA 0 EUR
APOSBIZY1029-1MG
Bacillus cereus Oligo-1,6-alpha-Glucosidase 13A
Enzymy
Oligo-1,6-alpha-Glucosidases participate in the hydrolysis of 1,6-alpha-Dglucosidic linkages in some oligosaccharides produced from starch and glycogen by alpha-amylases and isomaltose Family: GH13. E.C.:3.2.1.10. Architecture: GH13. Main Activity:oligosaccharide 1,6-alpha-glucosidase