Tlačiť
Paenibacillus polymyxa beta-Glucosidase 1B
Dodávateľ: Apollo Scientific
APOSBIZY1038-1MGEA 0 EUR
APOSBIZY1038-1MG
Paenibacillus polymyxa beta-Glucosidase 1B
Enzymy
beta-Glucosidases are enzymes that participate in the hydrolysis of terminal, non-reducing beta-D-glucosyl residues with release of beta-D-glucose. Family: GH1. E.C.:3.2.1.21. Architecture: GH1. Main Activity:-1,4-beta-glucosidase