Tlačiť
Thermotoga petrophila beta-Glucosidase 1A
Dodávateľ: Apollo
APOSBIZY1042-1MGEA 258.25 EUR
APOSBIZY1042-1MG
Thermotoga petrophila beta-Glucosidase 1A
Enzymy
beta-Glucosidases are enzymes that participate in the hydrolysis of terminal, non-reducing beta-D-glucosyl residues with release of beta-D-glucose. Family: GH1. E.C.:3.2.1.21. Architecture: GH1. Main Activity:-1,4-beta-glucosidase