Tlačiť
Thermotoga maritima alpha-glucuronidase 4A
Dodávateľ: Apollo Scientific
APOSBIZY1051-250UGEA 0 EUR
APOSBIZY1051-250UG
Thermotoga maritima alpha-glucuronidase 4A
Enzymy
Glucuronidases are enzymes that act on alpha- or beta-D-glucuronosides to release D-glucuronic acid residues. Family: GH4. E.C.:3.2.1.139. Architecture: GH4. Main Activity:-alpha-glucuronidase