Tlačiť
Bacteroides ovatus alpha-Methyl-glucuronidase 115A
Dodávateľ: Apollo
APOSBIZY1053-1MGEA 258.25 EUR
APOSBIZY1053-1MG
Bacteroides ovatus alpha-Methyl-glucuronidase 115A
Enzymy
Glucuronidases are enzymes that act on alpha- or beta-D-glucuronosides to release D-glucuronic acid residues. Family: GH115. E.C.:3.2.1.-. Architecture: GH115. Main Activity:-alpha-(4-O-methyl)-glucuronidase