Tlačiť
Ultrasonic baths, Elmasonic Easy
Dodávateľ: Elma
Elmasonic
ELMA1071673EA 1518.51 EUR
ELMA1071673 ELMA1071648 ELMA1071666 ELMA1071669 ELMA1071646 ELMA1071664 ELMA1071655 ELMA1071656 ELMA1071657 ELMA1071652 ELMA1071675 ELMA1071671
Ultrasonic baths, Elmasonic Easy
Lázně ultrazvukové
The Elmasonic EASY series of ultrasonic baths include a range of 9 different device sizes with tank volumes ranging from 0,8 to 28 litres.
  • Efficient 37 kHz ultrasonic high-performance transducers
  • Cleaning tank made from cavitation-resistant stainless steel
  • User-friendly and clearly arranged operating panel; sealed against liquid intrusion to protect the electronics
  • Continuous operating or short-term operation between 1 and 30 minutes
  • Permanent sweep function for optimised sound field distribution through frequency modulation
  • Temperature is selected via rotary switch; from 30 to 80 °C in increments of 5 °C (for H units)
  • Automatic safety shutdown after 8 hours of continuous operation or temperature of 90 °C
Microprocessor-controlled ultrasonic baths with sweep technology offer a state-of-the-art solution.
Informace o dodávce: Supplied with cover.