Tlačiť
Agarose LM, molecular biology grade, Axygen®
Dodávateľ: Corning Life Sciences
AXYGAGR-LM-1001EA 8440.88 EUR
AXYGAGR-LM-1001
Agarose LM, molecular biology grade, Axygen®
Gely elektroforetické Agaróza a Akrylamid
Axygen® Agarose Low Melting (LM) is a low melting agarose that allows for the recovery of undamaged nucleic acids below their denaturation temperature.