Tlačiť
Axygen® Axypet™ Pro single channel pipettors
Dodávateľ: Corning
Axygen® Axypet™
AXYGAP-20-PEA 330.32 EUR
AXYGAP-20-P AXYGAP-10-P AXYGAP-100-P AXYGAP-200-P AXYGAP-1000-P AXYGAP-50-P
Axygen® Axypet™ Pro single channel pipettors
Pipety Micropipettes
The Axypet™ pro pipettors have been engineered to provide the highest levels of comfort, accuracy and precision. Lightweight construction, contoured shape of the handle and four digit counter were designed to guarantee comfortable pipetting. All pipettors feature smooth plunger movement and extremely low pipetting forces to reduce the wrist strain and fatigue (RSI). Color-coded push buttons help to identify the size of the pipettor.
  • Ergonomic handle design for perfect fit for both right and left hand
  • Convenient one-hand volume setting with auto-lock system prevents accidental volume change during work
  • Four digit counter for greater precision, perfectly positioned for visibility at all times
  • Single-channel pipettors are compatible with very narrow tubes
  • Color-coded for easy size identification
  • Fully autoclavable and UV-resistant for sample protection against contaminations
  • Easy in-lab calibration helps you quickly adjust your pipettor to various liquid types
  • Individually tested and supplied with a certificate of quality
  • Three year manufacturer warranty
Certifikácie: Calibration certificate under ISO8655.