Tlačiť
Axygen® Axypet® Pro Multi-channel pipettors
Dodávateľ: Corning
Axygen® Axypet™
AXYGAP-12-300-PEA 872.81 EUR
AXYGAP-12-300-P AXYGAP-8-50-P AXYGAP-8-200-P AXYGAP-12-200-P AXYGAP-12-10-P AXYGAP-8-300-P AXYGAP-8-10-P AXYGAP-12-50-P
Axygen® Axypet® Pro Multi-channel pipettors
Pipety Micropipettes
The Axypet® pro pipettors have been engineered to provide the highest levels of comfort, accuracy and precision. Lightweight construction, contoured shape of the handle and four digit counter were designed to guarantee comfortable pipetting. All pipettors feature smooth plunger movement and extremely low pipetting forces to reduce the wrist strain and fatigue (RSI). Colour-coded push buttons help to identify the size of the pipettor.
  • Ergonomic handle design for perfect fit for both right and left hand
  • Fully autoclavable and UV-resistant for sample protection against contaminations
  • Easy in-lab calibration helps you quickly adjust your pipettor to various liquid types
  • Individually tested and supplied with a certificate of quality
  • Three year manufacturer warranty
Convenient one-hand volume setting with auto-lock system prevents accidental volume change during work. Four digit counter for greater precision, perfectly positioned for visibility at all times.The multi-channel shafts retract individually for perfect tip loading and easier tip ejection.
Certifikácie: Calibration certificate under ISO8655.