Tlačiť
Horizontal gel box, 15 cm, Axygen®
Dodávateľ: Corning
AXYGHGB-15EA 903.8 EUR
AXYGHGB-15
Horizontal gel box, 15 cm, Axygen®
Systémy elektroforetické Systémy pro horizontální gelovou elektroforézu
The Axygen HGB-15 is large sample capacity system ideal for screening PCR samples or routine DNA fragment analysis and comes standard with 2 different size UV transparent gel trays providing flexibility and value
The one piece injection molded tank is durable and leak free while the study electrode cassettes are user changeable. This compact unit is capable of separating bands over a distance of 15cm. The system only requires 500ml of buffer making it an economical choice.
System includes the novel Axygen contrast enhancer that allows for easy visualization of wells for sample loading and is reversible with white background to visualize running of gels with colored MW markers.
The external gel casting provides efficiency as gel casting and gel running can occur simultaneously.
Objednávacie informácie: Systems are supplied with manufacturer’s three-year warranty, and include a 15x10cm gel tray, a 15x15cm gel tray, rubber dams, Axygen contrast enhancer, 1 x 16MC well, 1.5mm thick and 1 x 20 well, 1.5mm thick combs , and 4mm power cables.