Tlačiť
Agarose MS, PCR screening, molecular biology grade, Axygen®
Dodávateľ: Corning Life Sciences
AXYGAGR-MS-1001EA 4572.53 EUR
AXYGAGR-MS-1001
Agarose MS, PCR screening, molecular biology grade, Axygen®
Gely elektroforetické Agaróza a Akrylamid
Axygen® Agarose Molecular Screening (MS) is a molecular screening agarose for improved resolution of DNA fragments with 500bp or less, especially primer-sized fragments.
  • Separation range - 1 to 2.5 kbp