Tlačiť
What is osmosis?
Dodávateľ: Edvotek
EDVOS-74EA 111.75 EUR
EDVOS-74
What is osmosis?
Using this kit, students will be able to see and understand the principles of osmosis. Using dialysis tubing, various salt concentrations, and dyes of different molecular weights it visually shows osmosis in action.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: 300 to 400 ml beakers, table salt, apple and beetroot juice, and distilled water.
Informace o dodávce: Kit includes instructions, high and low molecular weight dyes, dialysis tubing, and transfer pipettes.