Tlačiť
Chip ice makers
Dodávateľ: Ziegra Eismaschinen
474-0125 474-0126 474-0127 474-0149 474-0128 474-0070 474-0071 474-0072 474-0073 474-0074 474-0075 474-0153 474-0077 474-0078 474-0079 474-1245 474-0114 474-1225 474-0115 474-0116 474-0139 474-0117 474-0080 474-0081 474-0082 474-0083 474-1270 474-0084 474-0062 474-0085 474-0063 474-0064 474-0065 474-0121 474-0066 474-1210 474-0122 474-0067 474-0123 474-0068 474-0069 474-0124
Chip ice makers
Výrobníky ledu
Ice machines for small and moderate requirements with storage and other accessories, perfect for hospitals and laboratories. Ziegra ice has the ideal temperature of −0,5 °C, which means it never freezes together - even during prolonged storage, it always remains granular and free flowing. Chip ice is hygienic due to its closed water system and can be made from demineralised or tap water.
  • The static evaporator prevents refrigerant leaks
  • Different sizes of ice available; standard and micro (please state required size when ordering)
  • Compact constructrion and low noise emission
  • Automatic shut-down when the bin is full; automatic re-start when ice is removed
Two ice types available for every lab model:
Ziegra Standard-Ice melts slowly and is perfect for medical applications and preservation.
Ziegra Micro-Ice as an option, provides quick but gentle cooling to 0 °C, e.g. for test tubes or when added to liquids.
Cabinet and bin made of stainless steel.
Objednávacie informácie: Four models - ZBE 70, 150, 250 and 350 do not have integrated bins. These need to be ordered separately. See ordering table below for details.
Informace o dodávce: All installation fittings included.
Pozor: Ice storage bin must be ordered separately.