Tlačiť
Cryolys® Accessory for Precellys 24 and Evolution
Dodávateľ: Bertin
Cryolys®
432-3756 431-0230
Cryolys® Accessory for Precellys 24 and Evolution
Krabice Chladící/Mrazící boxy/Sáčky
Cryolys is a patented cooling option for Precellys homogenisers that prevents temperature increases during the homogenisation process, maintaining it within the physiological range, between 4 and 20 °C, depending on settings, strength and duration of homogenisation.
  • Cryolys prevents the temperature dependent degradation or denaturation of sensitive molecules like RNA or native proteins
  • Gives higher yields of intact target molecules during subsequent extraction and obtains better results during downstream analysis
Alternatively the Cryolys Evolution (432-0346) is available, which has been specifically designed for the Precellys Evolution homogeniser and only requires dry ice for operation.
Objednávacie informácie: The Cryolys models listed below require dry compressed air and liquid nitrogen or dry ice to operate.