Tlačiť

VWR® Trypsin

Dodávateľ: Avantor
VWR®
0785-5GEA 0 EUR
0785-5G
VWR® Trypsin
Enzymy
From bovine pancreas.

  • EINECS: 232-650-8
  • GHS Symbol 1: GHS08
  • GHS Symbol 2: GHS07
  • MDL: MFCD00082094

Product is tested to USP specifications.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR