Tlačiť
Nutrient media steriliser, MediaClave 30
Dodávateľ: Integra Biosciences
MediaClave
710-0736
Nutrient media steriliser, MediaClave 30
Příprava médií
The MediaClave 30 culture media steriliser for 3 to 30 L of nutrient media is a mobile, innovative device for gentle and reliable production of culture media with high-end requirements. It features state-of-the-art technology and is simple to operate, making it equally suited to standard types of agar and highly sensitive media. The efficient, innovative continuous-flow heater, powerful magnetic stirrer and rapid re-cooling process ensure that the media are sterilised safely and consistently at the required temperatures. The MediaClave 30 complies with the latest requirements for device documentation and of course conforms to GMP/GLP and FDA CFR21 Part 11. With an automatic cleaning program and highly efficient rapid re-cooling process.
  • 50 programs
  • Ethernet interface/WebServer
  • Precise control over temperature, pressure and time
  • Mobile and flexible
  • TÜV/CE certified
  • LCD display