Tlačiť
Spectrophotometers, Jenway® 7300 and 7305
Dodávateľ: Cole Parmer
664-0066EA 7459.69 EUR
664-0066 664-0067
Spectrophotometers, Jenway® 7300 and 7305
Spektrofotometry a fotometry Spektrofotometry UV/Vis
These spectrophotometers are designed to provide a compact instrument for use in education and quality check laboratories.
  • Icon driven software for easy and intuitive navigation
  • Small footprint with graphical display in the compartment lid
  • Autologging capabilities and optional internal printer
Informace o dodávce: Supplied fitted with 10×10 mm cuvette holder, 100 disposable cuvettes, instruction manual, power supply, PC software on CD ROM and interface cable.